Learn More
ACROBiosystems HRSV Post-Fusion Glycoprotein F0 Specific ELISA Kit, 96 Tests
Catalog No. NC3718200
Encompass_Preferred
A sandwich ELISA kit for specific, quantitative detection of human respiratory syncytial virus (HRSV) post-fusion glycoprotein F0 in vaccine samples. Designed for vaccine development, manufacturing, and quality control, the kit provides reliable measurement of post-fusion F0 antigen levels.
  • Sandwich ELISA format for quantitative detection.
  • Intended for vaccine development, manufacturing, and quality control.
  • Kit size: 96 tests.
  • Pre-coated antibody microplate (1 plate).
  • Post-fusion glycoprotein F0 standard, 20 μg.
  • HRP-conjugated anti-RSV F0 detection antibody, 20 μg.
  • 10x washing buffer, 50 mL; 2x dilution buffer, 50 mL.
  • Substrate solution, 12 mL; stop solution, 7 mL.
Catalog No. NC3718200 Supplier ACROBiosystems Supplier No. RASA20496TESTS
A sandwich ELISA kit for specific, quantitative detection of human respiratory syncytial virus (HRSV) post-fusion glycoprotein F0 in vaccine samples. Designed for vaccine development, manufacturing, and quality control, the kit provides reliable measurement of post-fusion F0 antigen levels.
  • Sandwich ELISA format for quantitative detection.
  • Intended for vaccine development, manufacturing, and quality control.
  • Kit size: 96 tests.
  • Pre-coated antibody microplate (1 plate).
  • Post-fusion glycoprotein F0 standard, 20 μg.
  • HRP-conjugated anti-RSV F0 detection antibody, 20 μg.
  • 10x washing buffer, 50 mL; 2x dilution buffer, 50 mL.
  • Substrate solution, 12 mL; stop solution, 7 mL.