Learn More
ACROBiosystems HRSV Pre-Fusion Glycoprotein F0 Specific ELISA Kit, 96 Tests
Catalog No. NC3718196
Encompass_Preferred
The HRSV Pre-Fusion Glycoprotein F0 Specific ELISA Kit is a sandwich ELISA designed for quantitative detection of HRSV pre-fusion F0 glycoprotein in vaccine samples. It is intended to support manufacturing and quality-control workflows by specifically identifying pre-fusion F0 from HRSV A and B while distinguishing it from post-fusion forms.
  • Kit provides 96 tests.
  • Sandwich ELISA method for quantitative detection.
  • Specifically detects HRSV (A) and HRSV (B) pre-fusion glycoprotein F0.
  • Does not detect post-fusion glycoprotein F0.
  • Intended for vaccine sample testing in manufacturing and quality control.
  • Optimized for quantitative determination in vaccine samples.
Catalog No. NC3718196 Supplier ACROBiosystems Supplier No. RASA20596TESTS
The HRSV Pre-Fusion Glycoprotein F0 Specific ELISA Kit is a sandwich ELISA designed for quantitative detection of HRSV pre-fusion F0 glycoprotein in vaccine samples. It is intended to support manufacturing and quality-control workflows by specifically identifying pre-fusion F0 from HRSV A and B while distinguishing it from post-fusion forms.
  • Kit provides 96 tests.
  • Sandwich ELISA method for quantitative detection.
  • Specifically detects HRSV (A) and HRSV (B) pre-fusion glycoprotein F0.
  • Does not detect post-fusion glycoprotein F0.
  • Intended for vaccine sample testing in manufacturing and quality control.
  • Optimized for quantitative determination in vaccine samples.