Learn More
ACROBiosystems ACROBiosystems HRV-3C Protease Cleavage Enzyme, GST Tag (active enzyme)
Catalog No. 502107421
Encompass_Preferred
Recombinant Protein;1000U;The enzyme recognizes the cleavage site: Leu-Glu-Val-Leu-Phe-Gln-↓-Gly-Pro. ;HRV-3C Protease Cleavage Enzyme, GST Tag is a recombinant form of human rhinovirus (HRV) type 14 3C protease produced in Escherichia coli cells at ACRObiosystems. ;This protein carries a GST tag at the N-terminus. The protein has a calculated MW of 46.0 kDa. The protein migrates as 44-46 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-210-7421 Supplier ACROBiosystems Supplier No. 3CCN31331000U
Recombinant Protein;1000U;The enzyme recognizes the cleavage site: Leu-Glu-Val-Leu-Phe-Gln-↓-Gly-Pro. ;HRV-3C Protease Cleavage Enzyme, GST Tag is a recombinant form of human rhinovirus (HRV) type 14 3C protease produced in Escherichia coli cells at ACRObiosystems. ;This protein carries a GST tag at the N-terminus. The protein has a calculated MW of 46.0 kDa. The protein migrates as 44-46 kDa under reducing (R) condition (SDS-PAGE).