Learn More
Sigma Aldrich Fine Chemicals Biosciences HRV 3C Protease. N Termina1MG
Catalog No. 502905599
Encompass
HRV-3C protease from human rhinovirus type 14 is a protease that specifically cleaves within an eight-residue recognition sequence.Proteolytic cleavage occurs between the Gln and Gly residues.HRV-3C protease is a useful tool to cleave recombinant proteins that are expressed as a fusion protein with this sequence between the carrier domain and the protein of interest.This recombinant version contains a six-histidine tag and can be easily removed by IMAC chromatography.
Catalog No. 50-290-5599 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SAE00451MG
HRV-3C protease from human rhinovirus type 14 is a protease that specifically cleaves within an eight-residue recognition sequence.Proteolytic cleavage occurs between the Gln and Gly residues.HRV-3C protease is a useful tool to cleave recombinant proteins that are expressed as a fusion protein with this sequence between the carrier domain and the protein of interest.This recombinant version contains a six-histidine tag and can be easily removed by IMAC chromatography.