Learn More
Perkin Elmer US LLC Wide-Bore Stainless Steel Replacement Needle
Catalog No. 509042214
Encompass_Preferred
Trap supplies from us, the market leader in thermal desorption, will provide exceptional analytical performance. Used for U.S. EPA Method TO17, our standard trap, packed with Tenax™, on the TurboMatrix™ Thermal Desorber, will improve productivity and trapping capacity. The TurboMatrix air monitoring trap is packed with carbonaceous sorbents suitable for ozone precursor and air toxics monitoring.
Catalog No. 50-904-2214 Supplier Perkin Elmer US LLC Supplier No. B0131385
Trap supplies from us, the market leader in thermal desorption, will provide exceptional analytical performance. Used for U.S. EPA Method TO17, our standard trap, packed with Tenax™, on the TurboMatrix™ Thermal Desorber, will improve productivity and trapping capacity. The TurboMatrix air monitoring trap is packed with carbonaceous sorbents suitable for ozone precursor and air toxics monitoring.