Learn More
Medchemexpress LLC HY-112823B 10mg Medchemexpress, HS-10296 hydrochloride CAS:2134096-03-8 Purity:>98%
SDP
Catalog No. 501969185
Encompass_Preferred
Medchemexpress, HY-112823B 10mg HS-10296 HCl CAS:2134096-03-8 HS-10296 hydrochloride is an orally available and third-Generation inhibitor of epidermal growth factor receptor (EGFR)-activating mutations and T790M-resistant mutation with limited activity against wild-type EGFR. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9185 Supplier Medchemexpress LLC Supplier No. HY112823B10MG
Medchemexpress, HY-112823B 10mg HS-10296 HCl CAS:2134096-03-8 HS-10296 hydrochloride is an orally available and third-Generation inhibitor of epidermal growth factor receptor (EGFR)-activating mutations and T790M-resistant mutation with limited activity against wild-type EGFR. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.