Learn More
Medchemexpress LLC HY-114349 10mg Medchemexpress, HS-1371 CAS:2158197-70-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11434910MG
Encompass_Preferred
Medchemexpress, HY-114349 10mg HS-1371 CAS:2158197-70-5 HS-1371 is a potent and ATP-competitive receptor-interacting protein kinase 3 (RIP3) inhibitor with an IC50 of 20.8 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9333