Learn More
TARGETMOL CHEMICALS INC HS-173 10MG
SDP
Catalog No. 502577167
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HS-173 is an effective PI3K(alpha) inhibitor (IC50 0.8 nM). purity: 99%
Catalog No. 50-257-7167 Supplier TARGETMOL CHEMICALS INC Supplier No. T230810MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HS-173 is an effective PI3K(alpha) inhibitor (IC50 0.8 nM). purity: 99%