Learn More
Medchemexpress LLC HY-15868 50mg , HS-173 CAS:1276110-06-5 Purity:98%
SDP
Catalog No. 501493967
Encompass_Preferred
Medchemexpress, HY-15868 50mg HS-173 CAS:1276110-06-5 Formula:C21H18N4O4S IC50: 0.8 nM (PI3Kα) Purity:98% HS-173 is a novel PI3K inhibitor, that is used for cancer treatment. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3967 Supplier Medchemexpress LLC Supplier No. HY1586850MG
Medchemexpress, HY-15868 50mg HS-173 CAS:1276110-06-5 Formula:C21H18N4O4S IC50: 0.8 nM (PI3Kα) Purity:98% HS-173 is a novel PI3K inhibitor, that is used for cancer treatment. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.