Learn More
TARGETMOL CHEMICALS INC HS-243 5MG
SDP
Catalog No. 502579703
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. HS-243 is an inhibitor of IRAK-4 and IRAK-1 with IC50s of 20 and 24 nM. HS-243 shows anti-inflammatory and anticancer activity. purity: 97%
Catalog No. 50-257-9703 Supplier TARGETMOL CHEMICALS INC Supplier No. T600125MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. HS-243 is an inhibitor of IRAK-4 and IRAK-1 with IC50s of 20 and 24 nM. HS-243 shows anti-inflammatory and anticancer activity. purity: 97%