Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HS-243 | 848249-10-5 | 100.0% | C17H16N4O3 | 10MG
SDP

Catalog No. 5000261373
Encompass_Preferred

HS-243 is a small-molecule research inhibitor of interleukin-1 receptor-associated kinases (IRAKs) with demonstrated anti-inflammatory and anticancer activity. It potently inhibits IRAK-4 (IC50 20 nM) and IRAK-1 (IC50 24 nM), and shows limited off-target activity against TAK1. Supplied for research use only.

  • Potent inhibition of IRAK-4 and IRAK-1 (IC50: 20 nM and 24 nM).
  • Limited TAK1 inhibition (IC50: ~0.5 μM).
  • High reported purity (99.95%).
  • Available as solid and DMSO solution formats.
  • Suitable for preclinical anti-inflammatory and anticancer studies.

Catalog No. 50-002-61373 Supplier Medchemexpress LLC Supplier No. HY13491110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HS-243 is a small-molecule research inhibitor of interleukin-1 receptor-associated kinases (IRAKs) with demonstrated anti-inflammatory and anticancer activity. It potently inhibits IRAK-4 (IC50 20 nM) and IRAK-1 (IC50 24 nM), and shows limited off-target activity against TAK1. Supplied for research use only.

  • Potent inhibition of IRAK-4 and IRAK-1 (IC50: 20 nM and 24 nM).
  • Limited TAK1 inhibition (IC50: ~0.5 μM).
  • High reported purity (99.95%).
  • Available as solid and DMSO solution formats.
  • Suitable for preclinical anti-inflammatory and anticancer studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.