Learn More
Medchemexpress LLC HS-27 | 1562024-11-6 | 98.5% | 1 ML
SDP
Catalog No. 5000340261
Encompass_Preferred
HS-27 is a fluorescently-tethered Hsp90 inhibitor that assays surface Hsp90 expression on intact tissue specimens. It is composed of the core elements of SNX-5422, an Hsp90 inhibitor, linked via a PEG linker to a fluorescein derivative (fluorescein isothiocyanate or FITC). This compound binds to ectopically expressed Hsp90 and has potential use in a "see-and-treat" paradigm in breast cancer.
  • Labels all receptor subtypes of breast cancer cells, but not normal cells
  • Specifically binds to Hsp90 expressed on the surface of breast cancer cells before being internalized
  • Fluorescence is greater in tumor than non-tumor tissue
Catalog No. 50-003-40261 Supplier Medchemexpress LLC Supplier No. HY13085110MM/1ML
HS-27 is a fluorescently-tethered Hsp90 inhibitor that assays surface Hsp90 expression on intact tissue specimens. It is composed of the core elements of SNX-5422, an Hsp90 inhibitor, linked via a PEG linker to a fluorescein derivative (fluorescein isothiocyanate or FITC). This compound binds to ectopically expressed Hsp90 and has potential use in a "see-and-treat" paradigm in breast cancer.
  • Labels all receptor subtypes of breast cancer cells, but not normal cells
  • Specifically binds to Hsp90 expressed on the surface of breast cancer cells before being internalized
  • Fluorescence is greater in tumor than non-tumor tissue