Learn More
Medchemexpress LLC Hs-276 | 2767422-72-8 | 98.9% | C24H29N5O2 | 10MG
SDP
Catalog No. 5000258879
Encompass_Preferred
HS-276 is an orally active, potent, and highly selective TAK1 inhibitor designed for laboratory research. It exhibits a reported Ki of 2.5 nM and is provided as a high-purity reagent for use in in vitro and preclinical studies focused on kinase signaling and inflammatory pathways.
  • Potent TAK1 inhibition with reported Ki = 2.5 nM.
  • Orally active small molecule for preclinical research.
  • High purity (98.9%) suitable for biological assays.
  • Molecular formula C24H29N5O2; molecular weight 419.52.
  • Available in multiple pack sizes for experimental flexibility.
Catalog No. 50-002-58879 Supplier Medchemexpress LLC Supplier No. HY14714110MG
HS-276 is an orally active, potent, and highly selective TAK1 inhibitor designed for laboratory research. It exhibits a reported Ki of 2.5 nM and is provided as a high-purity reagent for use in in vitro and preclinical studies focused on kinase signaling and inflammatory pathways.
  • Potent TAK1 inhibition with reported Ki = 2.5 nM.
  • Orally active small molecule for preclinical research.
  • High purity (98.9%) suitable for biological assays.
  • Molecular formula C24H29N5O2; molecular weight 419.52.
  • Available in multiple pack sizes for experimental flexibility.