Learn More
Selleck Chemical LLC HS-276-E1155-5MG
SDP
Catalog No. 503134440
Encompass_Preferred
HS-276 a potent and highly selective orally bioavailable transforming growth factor- activated kinase-1 (TAK1) inhibitor with the Ki and IC50 of 2 5 nM and 2 3 nM respectively shows significant inhibition of TNF-mediated cytokine profiles correlating with significant attenuation of arthritic-like symptoms in the collagen-induced arthritis (CIA) mouse model of inflammatory rheumatoid arthritis (RA)
Catalog No. 50-313-4440 Supplier Selleck Chemical LLC Supplier No. E11555MG
HS-276 a potent and highly selective orally bioavailable transforming growth factor- activated kinase-1 (TAK1) inhibitor with the Ki and IC50 of 2 5 nM and 2 3 nM respectively shows significant inhibition of TNF-mediated cytokine profiles correlating with significant attenuation of arthritic-like symptoms in the collagen-induced arthritis (CIA) mouse model of inflammatory rheumatoid arthritis (RA)