Learn More
Perkin Elmer US LLC Stainless Steel Needle, Jet, TurboMatrix 40 Replacement Parts
Supplier:  Perkin Elmer US LLC B4000011
Encompass_Preferred
TurboMatrix Headspace and high-sensitivity Headspace Trap samplers provide unparalleled precision and ease of use for numerous GC or GC/MS volatile-analysis applications. The system can manage up to 12 samples simultaneously, ensuring that the next sample is ready for analysis upon completion of the previous run, achieving significant time savings.
Catalog No. 50-904-2882