Learn More
ATCC Hs 729T; Rhabdomyosarcoma; Human (Homo sapiens)
Supplier:  ATCC HTB153
Encompass
Hs 729 [Hs 729T] is a fibroblast cell that was isolated from the muscle of a 74-year-old, White, male with rhabdomyosarcoma.
Catalog No. 50-238-2483