Learn More
ATCC Hs 895.Sk; Skin; Human (Homo sapiens)
Supplier:  ATCC CRL7636
Encompass
Hs 895.Sk are fibroblasts that were isolated from the skin of a normal 48-year-old, white female donor. It has applications in cancer research.
Catalog No. 50-238-4292