Learn More
ATCC Hs 940.T; Melanoma; Human (Homo sapiens)
Supplier:  ATCC CRL7691
Encompass
Hs 940.T are fibroblasts that were isolated from the skin of a 57-year-old, white male patient with melanoma. It has applications in cancer research.
Catalog No. 50-238-4410