Learn More
WELCH MATERIALS INC Ultisil HS-C18, 10µm, 120Å, 1kg
Catalog No. NC2052744
Encompass_Preferred
Bulk Media, Ultisil HS-C18, 10μm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 17%, Surface Area: 320m2/g, endcapped. Packing: 1kg/bottle.
Catalog No. NC2052744 Supplier WELCH MATERIALS INC Supplier No. H0272516100
Add to Cart
Add to Cart
Bulk Media, Ultisil HS-C18, 10μm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 17%, Surface Area: 320m2/g, endcapped. Packing: 1kg/bottle.