Learn More
YMC America, Inc HS C18 120A 5um 150x.3mm 1/16
SDP
Catalog No. 5000877282
Encompass
Hydrosphere C18 is based on highly inert ultra-pure pH neutral silica and is designed to maintain a hydrophilic surface for superior separation of hydrophilic compounds Minimal metal content and very low concentrations of residual silanols make this an ideal phase for the analysis of basic pharmaceuticals and chelating compounds even under LC-MS conditions such as separation in non-phosphate buffered systems In addition the ability to separate polar compounds under highly aqueous mobile phase conditions without loss in retention plate counts and peak performance greatly enhances the versatility of Hydrosphere C18
Catalog No. 50-008-77282 Supplier YMC America, Inc Supplier No. HS12S0515H0AU
Hydrosphere C18 is based on highly inert ultra-pure pH neutral silica and is designed to maintain a hydrophilic surface for superior separation of hydrophilic compounds Minimal metal content and very low concentrations of residual silanols make this an ideal phase for the analysis of basic pharmaceuticals and chelating compounds even under LC-MS conditions such as separation in non-phosphate buffered systems In addition the ability to separate polar compounds under highly aqueous mobile phase conditions without loss in retention plate counts and peak performance greatly enhances the versatility of Hydrosphere C18