Learn More
YMC America, Inc HS C18 col 120A 3um 20x2.1mm
SDP
Catalog No. 5000880961
Encompass_Preferred
Hydrosphere C18 is based on highly inert ultra-pure pH neutral silica and is designed to maintain a hydrophilic surface for superior separation of hydrophilic compounds Minimal metal content and very low concentrations of residual silanols make this an ideal phase for the analysis of basic pharmaceuticals and chelating compounds even under LC-MS conditions such as separation in non-phosphate buffered systems In addition the ability to separate polar compounds under highly aqueous mobile phase conditions without loss in retention plate counts and peak performance greatly enhances the versatility of Hydrosphere C18
Catalog No. 50-008-80961 Supplier YMC America, Inc Supplier No. HS12S0302Q1WT
Hydrosphere C18 is based on highly inert ultra-pure pH neutral silica and is designed to maintain a hydrophilic surface for superior separation of hydrophilic compounds Minimal metal content and very low concentrations of residual silanols make this an ideal phase for the analysis of basic pharmaceuticals and chelating compounds even under LC-MS conditions such as separation in non-phosphate buffered systems In addition the ability to separate polar compounds under highly aqueous mobile phase conditions without loss in retention plate counts and peak performance greatly enhances the versatility of Hydrosphere C18