Learn More
Trajan Scientific Headspace Cap Crimp 3mm, 5mm hole, 20mm Blue Silicone/Nat PTFE, 100 Pack
Catalog No. 50635103
Encompass_Preferred
L-MARK Brand Cap Crimp 20mm 3.0mm Blue Silicone/Nat PTFE Standard100 Pack
Catalog No. 50-635-103 Supplier Trajan Scientific Supplier No. XAR20038099
L-MARK Brand Cap Crimp 20mm 3.0mm Blue Silicone/Nat PTFE Standard100 Pack