Learn More
MyBioSource Mouse High Sensitivity C-Reactive Protein ELISA Kit

Supplier: MyBioSource MBS02698796STRIPWELLS
Short Name: High Sensitivity C-Reactive Protein
Reactivity: Mouse
Samples: Serum, Plasma, Cell Culture Supernatants, Tissue Homogenate, Urine and other Body Fluids
Detection Range: 6.25 ng/ml ~ 200 ng/ml.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.