Learn More
Crystal Chem Inc High Sensitivity C-Reactive Protein (hs-CRP) ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80955
Encompass
The High Sensitivity C-Reactive Protein ELISA is used to quantify levels of human c-reactive protein (CRP) in serum using only a 5 µL sample volume. Highlights: Kit uses sample size of only 5 µL. High sensitivity of 10 ng/mL)Wide dynamic range of 100 - 10000 ng/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7861