Learn More
Enzo Life Sciences AMPINEXT™ High-Sensitivity DNA Library Preparation Kit (Illumina) (12Reactions)
SDP
Catalog No. 502013789
Encompass_Preferred
The AMPINEXT™ High-Sensitivity DNA Library Preparation Kit (Illumina) is a complete set of optimized reagents to prepare a DNA library from very small amount of samples for use in next-generation sequencing applications. This kit is suitable for preparing a DNA library using sub-nanogram amounts of DNA input for next generation sequencing applications using an Illumina sequencer. These applications include genomic DNA-seq, ChIP-seq, MeDIP/hMeDIP-seq, traditional bisulfite-seq, and targeted re-sequencing. The optimized protocol and components of the kit allow both non-barcoded (singleplexed) and barcoded (multiplexed) DNA libraries to be constructed quickly with reduced bias. Assay Time: 1 hour 30 minutes.
Catalog No. 50-201-3789 Supplier Enzo Life Sciences Supplier No. ENZGEN5050012
The AMPINEXT™ High-Sensitivity DNA Library Preparation Kit (Illumina) is a complete set of optimized reagents to prepare a DNA library from very small amount of samples for use in next-generation sequencing applications. This kit is suitable for preparing a DNA library using sub-nanogram amounts of DNA input for next generation sequencing applications using an Illumina sequencer. These applications include genomic DNA-seq, ChIP-seq, MeDIP/hMeDIP-seq, traditional bisulfite-seq, and targeted re-sequencing. The optimized protocol and components of the kit allow both non-barcoded (singleplexed) and barcoded (multiplexed) DNA libraries to be constructed quickly with reduced bias. Assay Time: 1 hour 30 minutes.