Learn More
Agilent Technologies HS Large Fragment 50KB Kit, 500 samples, DNA Sizing Range: 75 bp - 50000 bp, DNA Sizing Accuracy: ± 15%, DNA Concentration Range: 5 pg/μL - 600 pg/μL, DNA Smear Range: 50 pg/μL - 5 ng/μL, Sample Volume Required: 2 μL
Catalog No. NC2080854
Encompass
HS Large Fragment 50KB Kit, 500 samples, DNA Sizing Range: 75 bp - 50000 bp, DNA Sizing Accuracy: ± 15%, DNA Concentration Range: 5 pg/μL - 600 pg/μL, DNA Smear Range: 50 pg/μL - 5 ng/μL, Sample Volume Required: 2 μL
Catalog No. NC2080854 Supplier Agilent Technologies Supplier No. DNF4640500
HS Large Fragment 50KB Kit, 500 samples, DNA Sizing Range: 75 bp - 50000 bp, DNA Sizing Accuracy: ± 15%, DNA Concentration Range: 5 pg/μL - 600 pg/μL, DNA Smear Range: 50 pg/μL - 5 ng/μL, Sample Volume Required: 2 μL