Learn More
Trajan Scientific Headspace Magnetic Crimp Cap 20mm, 5mm hole, PTFE/Clear Silicone, 3mm, 100 Pack
Catalog No. 50635111
Encompass_Preferred
L-MARK Brand Cap Crimp 20mm 3.0mm Blue Silicone/White PTFE Magnetic With 5mm hole100 Pack
Catalog No. 50-635-111 Supplier Trajan Scientific Supplier No. XAR20038331
L-MARK Brand Cap Crimp 20mm 3.0mm Blue Silicone/White PTFE Magnetic With 5mm hole100 Pack