Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent Technologies HS NGS Diluent Marker, 1 and 6000 bp, 2.4 mL, 1 Vial

Catalog No. NC1877468
Encompass

This analytical reference material is intended for use with Fragment Analyzer systems and the DNF-474 kit for NGS library preparation. The product is manufactured and verified in accordance with an ISO 9001:2015 certified quality assurance system, with characteristics tested on a per lot basis.

  • Assay type: NGS
  • Marker size: 1 bp and 6,000 bp
  • Storage temperature: -20°C
  • Volume: 2.4 mL
  • Pack type: 1 vial

Catalog No. NC1877468 Supplier Agilent Technologies Supplier No. DNF3730003
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This analytical reference material is intended for use with Fragment Analyzer systems and the DNF-474 kit for NGS library preparation. The product is manufactured and verified in accordance with an ISO 9001:2015 certified quality assurance system, with characteristics tested on a per lot basis.

  • Assay type: NGS
  • Marker size: 1 bp and 6,000 bp
  • Storage temperature: -20°C
  • Volume: 2.4 mL
  • Pack type: 1 vial

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.