Learn More
Medchemexpress LLC Hs-peg-sh (mw 3400) | 97.0% | 100 MG
SDP
Catalog No. 5000353960
Encompass_Preferred
Hs-peg-sh (mw 3400) is a linear homobifunctional PEG and a cross-linker. It is utilized in drug delivery and for the preparation of PEG hydrogels.
  • Linear homobifunctional PEG
  • Cross-linker
  • Used for drug delivery
  • Used for preparation of PEG hydrogels
Catalog No. 50-003-53960 Supplier Medchemexpress LLC Supplier No. HY145485100MG
Hs-peg-sh (mw 3400) is a linear homobifunctional PEG and a cross-linker. It is utilized in drug delivery and for the preparation of PEG hydrogels.
  • Linear homobifunctional PEG
  • Cross-linker
  • Used for drug delivery
  • Used for preparation of PEG hydrogels