Learn More
Medchemexpress LLC Hs-peg-sh (mw 3400) | 97.0% | 25 MG
SDP
Catalog No. 5000353267
Encompass_Preferred
HS-PEG-SH (MW 3400) is a linear homobifunctional PEG and a cross-linker, useful for drug delivery and the preparation of PEG hydrogels. It has a molecular weight of 3400 (Average) and appears as a white to light yellow solid. This product is for research use only.
  • Linear homobifunctional PEG
  • Cross-linker
  • Utilized for drug delivery
  • Used in the preparation of PEG hydrogels
  • White to light yellow solid appearance
  • Research use only
Catalog No. 50-003-53267 Supplier Medchemexpress LLC Supplier No. 25MGHY145485
HS-PEG-SH (MW 3400) is a linear homobifunctional PEG and a cross-linker, useful for drug delivery and the preparation of PEG hydrogels. It has a molecular weight of 3400 (Average) and appears as a white to light yellow solid. This product is for research use only.
  • Linear homobifunctional PEG
  • Cross-linker
  • Utilized for drug delivery
  • Used in the preparation of PEG hydrogels
  • White to light yellow solid appearance
  • Research use only