Learn More
Medchemexpress LLC HS-PEG-SH (MW 3400) | 97.0% | 50 MG
SDP
Catalog No. 5000353961
Encompass_Preferred
HS-PEG-SH (MW 3400) is a linear homobifunctional PEG cross-linker designed for various biochemical applications. It is utilized in drug delivery systems and for the preparation of PEG hydrogels, offering versatility for research purposes. This product has an average molecular weight of 3400 and a purity of 97.0%.
  • Linear homobifunctional PEG
  • Cross-linker for biochemical applications
  • Utilized for drug delivery
  • Suitable for preparing PEG hydrogels
  • 97.0% purity
  • Average molecular weight of 3400
  • Appears as a solid
  • Color ranges from white to light yellow
Catalog No. 50-003-53961 Supplier Medchemexpress LLC Supplier No. HY14548550MG
HS-PEG-SH (MW 3400) is a linear homobifunctional PEG cross-linker designed for various biochemical applications. It is utilized in drug delivery systems and for the preparation of PEG hydrogels, offering versatility for research purposes. This product has an average molecular weight of 3400 and a purity of 97.0%.
  • Linear homobifunctional PEG
  • Cross-linker for biochemical applications
  • Utilized for drug delivery
  • Suitable for preparing PEG hydrogels
  • 97.0% purity
  • Average molecular weight of 3400
  • Appears as a solid
  • Color ranges from white to light yellow