Learn More
Medchemexpress LLC HS-PEG3-CH2CH2NH2 hydrochloride | 2863676-86-0 | 95.0% | 100 MG
SDP
Catalog No. 5000341156
Encompass_Preferred
HS-PEG3-CH2CH2NH2 hydrochloride is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs utilize two distinct ligands connected by a linker
  • Selectively degrades target proteins through the ubiquitin-proteasome system
Catalog No. 50-003-41156 Supplier Medchemexpress LLC Supplier No. HY130871A100MG
HS-PEG3-CH2CH2NH2 hydrochloride is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs utilize two distinct ligands connected by a linker
  • Selectively degrades target proteins through the ubiquitin-proteasome system