Learn More
Medchemexpress LLC HS-PEG5-CH2CH2NH2 hydrochloride | 1446256-16-1 | 98.0% | 50 MG
SDP
Catalog No. 5000340286
Encompass_Preferred
HS-PEG5-CH2CH2NH2 hydrochloride, also known as Thiol-PEG5-amine hydrochloride, is a PROTAC linker belonging to the PEG class. It is utilized in the synthesis of PROTAC molecules.
  • It is a PROTAC linker.
  • It belongs to the PEG class of compounds.
  • It can be used to synthesize PROTAC molecules.
  • It has a molecular weight of 333.87.
  • Its appearance is a colorless to light yellow liquid.
  • It has a purity of 98.0%.
  • It is soluble in DMSO at concentrations of 250 mg/mL or greater.
  • For research use only; not for sale to patients.
Catalog No. 50-003-40286 Supplier Medchemexpress LLC Supplier No. HY130872A50MG
HS-PEG5-CH2CH2NH2 hydrochloride, also known as Thiol-PEG5-amine hydrochloride, is a PROTAC linker belonging to the PEG class. It is utilized in the synthesis of PROTAC molecules.
  • It is a PROTAC linker.
  • It belongs to the PEG class of compounds.
  • It can be used to synthesize PROTAC molecules.
  • It has a molecular weight of 333.87.
  • Its appearance is a colorless to light yellow liquid.
  • It has a purity of 98.0%.
  • It is soluble in DMSO at concentrations of 250 mg/mL or greater.
  • For research use only; not for sale to patients.