Learn More
Medchemexpress LLC HS-PEG5-CH2CH2NH2 hydrochloride | 1446256-16-1 | 98.0% | C12H28ClNO5S | 10MG
SDP
Catalog No. 5000261969
Encompass_Preferred
HS-PEG5-CH2CH2NH2 hydrochloride is a thiol-terminated PEG5 linker supplied as the hydrochloride salt, intended for synthesis of PROTACs and other bioconjugates. It provides a short, flexible spacer with terminal thiol and amine groups for versatile conjugation chemistries.
  • Thiol-terminated PEG5 linker for conjugation chemistries.
  • Terminal ethyleneamine available as hydrochloride salt.
  • Short, flexible spacer suitable for PROTAC design.
  • High purity (98.0%) for consistent synthetic performance.
  • Compatible with common bioconjugation reactions.
Catalog No. 50-002-61969 Supplier Medchemexpress LLC Supplier No. HY130872A10MG
HS-PEG5-CH2CH2NH2 hydrochloride is a thiol-terminated PEG5 linker supplied as the hydrochloride salt, intended for synthesis of PROTACs and other bioconjugates. It provides a short, flexible spacer with terminal thiol and amine groups for versatile conjugation chemistries.
  • Thiol-terminated PEG5 linker for conjugation chemistries.
  • Terminal ethyleneamine available as hydrochloride salt.
  • Short, flexible spacer suitable for PROTAC design.
  • High purity (98.0%) for consistent synthetic performance.
  • Compatible with common bioconjugation reactions.