Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HS-PEG6-CH2CH2-Boc | 1818294-40-4 | >98.0% | C19H38O8S | 10MG
SDP

Catalog No. 5000261812
Encompass_Preferred

HS-PEG6-CH2CH2-Boc is a polyethylene glycol (PEG)-based PROTAC linker featuring a protected thiol and a tert-butyloxycarbonyl (Boc)-protected terminus. It is supplied at high purity (>98%), has molecular formula C19H38O8S and molecular weight 426.57 g·mol⁻¹, and is soluble in DMSO (10 mM), making it suitable for use in small-molecule linker assembly.

  • Provides a flexible, hydrophilic spacer for connecting ligands in PROTAC synthesis.
  • Contains a protected thiol for selective deprotection and conjugation.
  • Features a Boc-protected terminus compatible with standard amine deprotection chemistry.
  • High reported purity supports reliable synthetic outcomes.
  • Soluble in DMSO at 10 mM for convenient handling in solution-phase reactions.

Catalog No. 50-002-61812 Supplier Medchemexpress LLC Supplier No. HY13088110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HS-PEG6-CH2CH2-Boc is a polyethylene glycol (PEG)-based PROTAC linker featuring a protected thiol and a tert-butyloxycarbonyl (Boc)-protected terminus. It is supplied at high purity (>98%), has molecular formula C19H38O8S and molecular weight 426.57 g·mol⁻¹, and is soluble in DMSO (10 mM), making it suitable for use in small-molecule linker assembly.

  • Provides a flexible, hydrophilic spacer for connecting ligands in PROTAC synthesis.
  • Contains a protected thiol for selective deprotection and conjugation.
  • Features a Boc-protected terminus compatible with standard amine deprotection chemistry.
  • High reported purity supports reliable synthetic outcomes.
  • Soluble in DMSO at 10 mM for convenient handling in solution-phase reactions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.