Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hs-peg7500-cooh

Catalog No. 503124895 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-4895 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-312-4895 Supplier Sigma Aldrich Supplier No. JKA51011G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Product is a thiolated carboxylic-polyethylene . 4α;-sulfanyl--d carboxy PEG (HS-PEG-COOH) heterobifunctional PEGs with functional end-groups of very different reactivity. HS-PEG-COOH inhibits nonspecific binding to living cells.HS-PEG-COOH may be tethered to the gold nanoparticle end during the fabrication of epidermal growth factor (EGF)-conjugated surface-enhanced Raman spectroscopy (SERS) nanoparticles. HS-PEG-COOH was coated onto silicon-on-insulator photonic biosensor chips. Quantum dots (QD)/ virus-like particles (VLPs) could be obtained only from HS-PEG-COOH-coated QDs. It may be used to stabilize nanoparticles and prevent its aggregation.Product of JenKem Technology

Specifications

Linear Formula HS(CH2CH2O)nCH2COOH
Quantity 1 g
Synonym HS-PEG-COOH; Thiol-PEG-Carboxyl, HS-PEG-COOH
Molecular Weight (g/mol) 7500
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.