Learn More
Sigma Aldrich Fine Chemicals Biosciences HS SelectR Nckel Afnt 500ML
Catalog No. 502914666
Encompass
HIS-Select(R) Nickel Affinity Gel is durable and can capture the recombinant proteins with histidine tags at a high flow rate. Recombinant proteins with histidine tags are bound using either native or denaturing conditions.
Catalog No. 50-291-4666 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P6611500ML
Add to Cart
Add to Cart
Request a Quote
HIS-Select(R) Nickel Affinity Gel is durable and can capture the recombinant proteins with histidine tags at a high flow rate. Recombinant proteins with histidine tags are bound using either native or denaturing conditions.