Learn More
Agilent Technologies HS Small Fragment Kit, 500
Catalog No. NC1768626
Encompass
Fragment Analyzer system kit for the quantitative and qualitative analysis of small DNA fragments and smears from 50 to 1,500 bp at low concentrations. Includes the reagents necessary for running 500 samples.
Catalog No. NC1768626 Supplier Agilent Technologies Supplier No. DNF4770500
Fragment Analyzer system kit for the quantitative and qualitative analysis of small DNA fragments and smears from 50 to 1,500 bp at low concentrations. Includes the reagents necessary for running 500 samples.