Learn More
Enzo Life Sciences AMPIGENE HS Taq DNA Polymerase (500U)
SDP
Catalog No. 502013898
Encompass_Preferred
AMPIGENE HS Taq DNA Polymerase uses the latest developments in polymerase technology and buffer chemistry to optimize and increase PCR sensitivity. The AMPIGENE HS Taq DNA Polymerase and 5x AMPIGENE reaction buffer are separate for flexibility and end-user customization. Hot-start technology is used to increase sensitivity for PCR applications including genotyping, multiplex PCR, screening, library construction, colony PCR and PCR direct from blood and urine. The AMPIGENE HS Taq DNA Polymerase is resistant to PCR inhibitors and suitable for direct PCR from unprocessed samples: bacterial culture, bacterial colonies, blood, and urine. Applications: PCR.
Catalog No. 50-201-3898 Supplier Enzo Life Sciences Supplier No. ENZPRT1010500
AMPIGENE HS Taq DNA Polymerase uses the latest developments in polymerase technology and buffer chemistry to optimize and increase PCR sensitivity. The AMPIGENE HS Taq DNA Polymerase and 5x AMPIGENE reaction buffer are separate for flexibility and end-user customization. Hot-start technology is used to increase sensitivity for PCR applications including genotyping, multiplex PCR, screening, library construction, colony PCR and PCR direct from blood and urine. The AMPIGENE HS Taq DNA Polymerase is resistant to PCR inhibitors and suitable for direct PCR from unprocessed samples: bacterial culture, bacterial colonies, blood, and urine. Applications: PCR.