Learn More
Enzo Life Sciences AMPIGENE HS Taq Mix (200Reactions)
SDP
Catalog No. 502013870
Encompass_Preferred
AMPIGENE HS Taq Mix uses the latest developments in polymerase technology and buffer chemistry to optimize and increase PCR sensitivity. The AMPIGENE HS Taq Mix is a robust mix using hot-start technology to increase sensitivity for PCR applications including genotyping, multiplex PCR, screening, library construction, colony PCR and PCR direct from blood and urine. An optimized buffer system allows efficient amplification under fast and standard cycling conditions. The AMPIGENE HS Taq Mix is resistant to PCR inhibitors and suitable for direct PCR from unprocessed samples: bacterial culture, bacterial colonies, blood, and urine. Applications: PCR.
Catalog No. 50-201-3870 Supplier Enzo Life Sciences Supplier No. ENZNUC1010200
AMPIGENE HS Taq Mix uses the latest developments in polymerase technology and buffer chemistry to optimize and increase PCR sensitivity. The AMPIGENE HS Taq Mix is a robust mix using hot-start technology to increase sensitivity for PCR applications including genotyping, multiplex PCR, screening, library construction, colony PCR and PCR direct from blood and urine. An optimized buffer system allows efficient amplification under fast and standard cycling conditions. The AMPIGENE HS Taq Mix is resistant to PCR inhibitors and suitable for direct PCR from unprocessed samples: bacterial culture, bacterial colonies, blood, and urine. Applications: PCR.