Learn More
As One International Inc Hot Start Taq Polymerase 5U/ul, 10x Ammonium Buffer and 10x Combination Buffer, 2,500U
Catalog No. 501655913
Encompass_Preferred
AS ONE HS DNA Polymerase is a modified form of AS ONE Taq DNA Polymerase, which is activated by heat treatment. A chemical moiety is attached to the enzyme at the active site, which renders the enzyme inactive at room temperature. Thus, during setup and the first ramp of thermal cycling, the enzyme is not active and misprimed primers are not extended. The result is higher specificity and greater yields when compared to standard DNA polymerases.    Once the reaction reaches optimal activating temperature, the chemical moiety is cleaved during a 15 minute heat activation step, releasing the active HS Polymerase DNA Polymerase into the reaction. HS Taq includes 10x Ammonium Buffer and 10x Combination Buffer
Catalog No. 50-165-5913 Supplier As One International Inc Supplier No. AO225106
AS ONE HS DNA Polymerase is a modified form of AS ONE Taq DNA Polymerase, which is activated by heat treatment. A chemical moiety is attached to the enzyme at the active site, which renders the enzyme inactive at room temperature. Thus, during setup and the first ramp of thermal cycling, the enzyme is not active and misprimed primers are not extended. The result is higher specificity and greater yields when compared to standard DNA polymerases.    Once the reaction reaches optimal activating temperature, the chemical moiety is cleaved during a 15 minute heat activation step, releasing the active HS Polymerase DNA Polymerase into the reaction. HS Taq includes 10x Ammonium Buffer and 10x Combination Buffer