Learn More
As One International Inc Hot Start Taq Polymerase 5U/ul, 10x Combination Buffer, 10,000U
Catalog No. 501655895
Encompass_Preferred
AS ONE HS Taq DNA Polymerase is a hot start polymerase designed to diminish the formation of non-specific priming events during reaction set up and the first ramp of thermal cycling. HS Taq features high specificity and sensitivity, greater yield compared to standard DNA polymerases. HS Taq enables the detection of low abundance tragets. HS Taq Polymerase includes 10x Combination Buffer.
Catalog No. 50-165-5895 Supplier As One International Inc Supplier No. AO223108
AS ONE HS Taq DNA Polymerase is a hot start polymerase designed to diminish the formation of non-specific priming events during reaction set up and the first ramp of thermal cycling. HS Taq features high specificity and sensitivity, greater yield compared to standard DNA polymerases. HS Taq enables the detection of low abundance tragets. HS Taq Polymerase includes 10x Combination Buffer.