Learn More
Cell Signaling Technology PTMScan HS Ubiquitin/SUMO Remnant Motif Kit, Enhanced Identification of Modified Peptides, High Sensitivity, 1 Kit
SDP
Catalog No. NC3251986
Encompass
The PTMScan HS Ubiquitin/SUMO Remnant Motif Kit from Cell Signaling Technology Inc. offers advanced identification and profiling of post-translationally modified peptides. Utilizing proprietary PTMScan technology, the kit enhances peptide enrichment through immunoprecipitation, followed by analysis using liquid chromatography tandem mass spectrometry (LC-MS/MS). This method provides high specificity and sensitivity, making it a valuable tool for researchers focusing on ubiquitin and SUMO remnant motifs.
Catalog No. NC3251986 Supplier Cell Signaling Technology Supplier No. 59322S
The PTMScan HS Ubiquitin/SUMO Remnant Motif Kit from Cell Signaling Technology Inc. offers advanced identification and profiling of post-translationally modified peptides. Utilizing proprietary PTMScan technology, the kit enhances peptide enrichment through immunoprecipitation, followed by analysis using liquid chromatography tandem mass spectrometry (LC-MS/MS). This method provides high specificity and sensitivity, making it a valuable tool for researchers focusing on ubiquitin and SUMO remnant motifs.