Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HS024 TFA | 99.8% | 1380.52 | 10 MG
SDP

Catalog No. 5000423488
Encompass_Preferred

HS024 is a selective MC4 receptor antagonist, with Kis of 0.29, 3.29, 5.45, and 18.6 nM for MC4, MC5, MC3, and MC1, respectively. HS024 increases food intake.

  • Selective MC4 receptor antagonist
  • Increases food intake
  • Molecular weight: 1380.52
  • Formula: C60H80F3N19O12S2
  • Appearance: Solid
  • Color: White to off-white
  • Sequence shortening: Ac-C-{Nle}-RH-{D-2Nal}-RWGC-NH2 (disulfide bridge: Cys1-Cys9)
  • Solubility (in vitro): H2O : 50 mg/mL (36.22 mM; need ultrasonic)

Catalog No. 50-004-23488 Supplier Medchemexpress LLC Supplier No. HYP1215A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HS024 is a selective MC4 receptor antagonist, with Kis of 0.29, 3.29, 5.45, and 18.6 nM for MC4, MC5, MC3, and MC1, respectively. HS024 increases food intake.

  • Selective MC4 receptor antagonist
  • Increases food intake
  • Molecular weight: 1380.52
  • Formula: C60H80F3N19O12S2
  • Appearance: Solid
  • Color: White to off-white
  • Sequence shortening: Ac-C-{Nle}-RH-{D-2Nal}-RWGC-NH2 (disulfide bridge: Cys1-Cys9)
  • Solubility (in vitro): H2O : 50 mg/mL (36.22 mM; need ultrasonic)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.