Learn More
Aviva Systems Biology Corp HS1BP3 Antibody - N-terminal region (ARP57633_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57633_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HS1BP3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HS1BP3. Alternate names: ETM2, FLJ14249, HS1-BP3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62917