Learn More
Aviva Systems Biology Corp HS2ST1 Antibody - middle region (ARP48741_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48741_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HS2ST1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HS2ST1. Alternate names: FLJ11317, KIAA0448, MGC131986, dJ604K5.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59909