Learn More
Aviva Systems Biology Corp HS3ST1 Antibody - middle region (ARP33092_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33092_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HS3ST1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HS3ST1. Alternate names: 3OST, 3OST1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54880