Learn More
Aviva Systems Biology Corp HS3ST3B1 Antibody - N-terminal region (ARP46685_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46685_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HS3ST3B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HS3ST3B1. Alternate names: 30ST3B1, 3OST3B1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-53483