Learn More
Aviva Systems Biology Corp HS3ST5 Antibody - C-terminal region (ARP49083_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49083_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HS3ST5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human HS3ST5. Alternate names: 3-OST-5, 3OST5, HS3OST5, NBLA04021ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53571