Learn More
Grace Bio Labs Inc HS6024 HYBRISLIP 60X24MM 100PK
Catalog No. NC9296662
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
HS6024-HybriSlip Hybridization Cover, 24mm X 60mm X 0.25mm Thickness - 100 PACK
Catalog No. NC9296662 Supplier Grace Bio Labs Inc Supplier No. 716024
This item has a minimum qty of 2 per supplier requirements.
HS6024-HybriSlip Hybridization Cover, 24mm X 60mm X 0.25mm Thickness - 100 PACK